Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hydroxide 33% (w/w) in water, extra pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

358620050 358620250 358625000
TS358620050EA 112 USD InStock
TS358620050 TS358620250 TS358625000
Sodium hydroxide 33% (w/w) in water, extra pure
Sodium hydroxide


Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 8 °C
Density: 1.36 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless Solution
Heavy metals ≤10 ppm
Density (20°C) 1.355 to 1.365 kg/l
Titration with HCl 32.5 to 33.5 %
Sodium carbonate (Na2CO3) ≤0.1 % (Na2CO3)
Sodium sulfate (Na2SO4) ≤50 ppm (Na2SO4)
Sodium chloride (NaCl) ≤50 ppm (NaCl)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty