Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium hydroxide 1 N in water

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

278400010 278400025 278400250
TS278400010EA 34 USD InStock
TS278400010 TS278400025 TS278400250
Potassium hydroxide 1 N in water
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 100 °C (760 mmHg)
Melting Pt: 360
Density: 1.05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806
Order Now
Specification Test Results
Appearance Clear colorless Solution
Strength 1 N Exact strength on the certificate of analysis

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty