Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-2-pyrrolidone (NMP) 98%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

449180050 449180250
TS449180050EA 166 USD InStock
TS449180050 TS449180250
N-Methyl-2-pyrrolidone (NMP) 98%, pure
N-Methyl-2-pyrrolidone (NMP)


Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.03 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
Merck Index: 13,06140
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4690 to 1.4710 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤100 APHA

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty