Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

p-Cresol 99+%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

405740050 405741000
TS405740050EA 29 USD InStock
TS405740050 TS405741000
p-Cresol 99+%, pure
p-Cresol


Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 202 °C (760 mmHg)
Melting Pt: 32…34 °C
Density: 1.034 g/cm³
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00002376
CAS Number: 106-44-5
EINECS: 203-398-6
UN: 3455
ADR: 6.1 II
Merck Index: 13,02604
Order Now
Specification Test Results
Appearance Colorless to light yellow Crystalline solid or liquid (May darken on storage)
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥99.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty