Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloroacetic acid 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

220320010 220321000
TS220320010EA 65 USD InStock
TS220320010 TS220321000
Chloroacetic acid 99+%
Chloroacetic acid


Formula: ClCH₂COOH
MW: 94.5 g/mol
Boiling Pt: 188
Melting Pt: 61…63 °C
Density: 1.58 g/cm³
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002683
CAS Number: 79-11-8
EINECS: 201-178-4
UN: 1751
ADR: 6.1 II
Merck Index: 13,02129
Order Now
Specification Test Results
Appearance White Crystals or flakes
Infrared spectrum Conforms
Melting point 61 to 63 °C
Water ≤0.3 %
Titration with NaOH ≥99.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty