Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloroacetic acid 99+% ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

448110025 448111000 448115000
TS448110025EA 242 USD InStock
TS448110025 TS448111000 TS448115000
Chloroacetic acid 99+% ACS
Chloroacetic acid


Formula: ClCH₂COOH
MW: 94.5 g/mol
Boiling Pt: 188
Melting Pt: 61…63 °C
Density: 1.58 g/cm³
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002683
CAS Number: 79-11-8
UN: 1751
ADR: 6.1 II
Merck Index: 13,02129
Order Now
Specification Test Results
Appearance Colorless to white Deliquescent crystals or flakes
Sulfated ash ≤0.02 %
Titration with NaOH ≥99.0 %
Substance darkened by H2SO4 passes test
Chloride (Cl) ≤0.01 %
Sulfate (SO4) ≤0.02 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.01 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty