Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perchloric acid 70% in water ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

424030010 424030025 424032500
TS424030010EA 218 USD InStock
TS424030010 TS424030025 TS424032500
Perchloric acid 70% in water ACS
Perchloric acid


Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 39
Melting Pt: -101
Density: 1.66 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1 I
Order Now
Specification Test Results
Appearance Clear liquid
Color scale ≤10 APHA
Residue after ignition ≤0.003 %
Titration with NaOH 69.0 to 72.0 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Silicate and phosphate ≤5 ppm (as SiO2)
Iron (Fe) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm
Nitrogen compounds (as N) ≤0.001 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty