Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trifluoroacetic acid-[D₁] (99.5% D) for NMR spectroscopy

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

325310100 325310250 325311000
TS325310100EA 91 USD InStock
TS325310100 TS325310250 TS325311000
Trifluoroacetic acid-[D₁] (99.5% D) for NMR spectroscopy
Trifluoroacetic acid-[D₁]


Formula: F₃CCOOD
MW: 115.02 g/mol
Boiling Pt: 75
Melting Pt: -15
Density: 1.5 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00044563
CAS Number: 599-00-8
EINECS: 209-961-2
UN: 2699
ADR: 8 I
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.05 % (Coulometric)
D-Enrichment ≥99.4 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty