Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gallic acid monohydrate ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

447581000 447585000
TS447581000EA 88 USD InStock
TS447581000 TS447585000
Gallic acid monohydrate ACS
Gallic acid monohydrate


Formula: C₇H₆O₅·1H₂O
MW: 188.14 g/mol
Melting Pt: 250 (dec.)
Flash Pt: 250
Storage Temperature: Ambient
MDL Number: MFCD00149098
CAS Number: 5995-86-8
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Residue after ignition ≤0.05 %
Titration with NaOH ≥98.0 %
Sulfate (SO4) ≤0.02
Insoluble matter ≤0.01 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty