Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Polyphosphoric acid (≥84% P₂O₅), pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

196950010 196950025
TS196950010EA 86 USD InStock
TS196950010 TS196950025
Polyphosphoric acid (≥84% P₂O₅), pure
Polyphosphoric acid


Formula: H(PO₃H)nOH
Boiling Pt: 550 °C (760 mmHg)
Density: 2.05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00084480
CAS Number: 8017-16-1
UN: 3264
ADR: 8 III
Order Now
Specification Test Results
Appearance Clear viscous liquid (May darken on storage)
Color scale ≤50 APHA
Titration with NaOH ≥82 % (as P2O5)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty