Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nitrobenzene-[D₅] 99 atom % D for NMR spectroscopy

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

166360050 166360100 166360250
TS166360050EA 61 USD InStock
TS166360050 TS166360100 TS166360250
Nitrobenzene-[D₅] 99 atom % D for NMR spectroscopy
Nitrobenzene-[D₅]


Formula: C₆D₅NO₂
Boiling Pt: 88 °C (12 mmHg)
Melting Pt: 5 °C
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00044415
CAS Number: 4165-60-0
EINECS: 224-014-3
UN: 1662
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Water ≤0.03 %
D-Enrichment ≥98.5 %
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty