Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Tartaric acid 99.5%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

137870025 137871000
TS137870025EA 397 USD InStock
TS137870025 TS137871000
DL-Tartaric acid 99.5%
DL-Tartaric acid


Formula: C₄H₆O₆
MW: 150.09 g/mol
Melting Pt: 200…206 °C
Flash Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00151254
CAS Number: 133-37-9
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 200 to 206 °C
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
Specific optical rotation -0.5° to +0.5° (20 °C, 589 nm) (c=20, H2O)
Titration with NaOH ≥99.4 %
Heavy metals (as Pb) ≤20 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty