Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

InVivoMAb Anti-NKG2D (CD314) Mouse Monoclonal Antibody [clone: 1D11]

Supplier: Bio X Cell
BE0351-1MG BE0351-25MG
77579-052EA 174.46 USD InStock
77579-052 77579-054
InVivoMAb Anti-NKG2D (CD314) Mouse Monoclonal Antibody [clone: 1D11]
Antibodies
Type: Primary
Antigen: NKG2D (CD314)
Clonality: Monoclonal
Clone: 1D11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty