Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

InVivoMAb Anti-CD2 Mouse Monoclonal Antibody [clone: OX-34]

Supplier: Bio X Cell
BE0417-100MG BE0417-1MG BE0417-25MG BE0417-50MG BE0417-5MG
77579-710EA 4408.4 USD InStock
77579-710 77579-712 77579-714 77579-716 77579-718
InVivoMAb Anti-CD2 Mouse Monoclonal Antibody [clone: OX-34]
Antibodies
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: OX-34
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty