Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nickel(II) iodide hexahydrate 98%

Supplier: AMBEED, INC
Danger

A983656-100G A983656-25G A983656-5G
77623-264EA 299.73 USD InStock
77623-264 77623-266 77623-268
Nickel(II) iodide hexahydrate 98%
Nickel(II) iodide hexahydrate
Precaución: for Research Use Only


Formula: H₁₂I₂NiO₆
MW: 420.59 g/mol
MDL Number: MFCD00150266
CAS Number: 7790-34-3
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty