Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-11-Methyl-2-dodecenoic acid 95%, Ambeed.Inc

Supplier: AMBEED, INC
Danger

A598084-10MG A598084-50MG
77602-490EA 95.33 USD InStock
77602-490 77684-469
cis-11-Methyl-2-dodecenoic acid 95%, Ambeed.Inc
cis-11-Methyl-2-dodecenoic acid
Precaución: For research use only.


Formula: C₁₃H₂₄O₂
MDL Number: MFCD09263531
CAS Number: 677354-23-3
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty