Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Decyl-3-methylimidazoliumtetrafluoroborate ≥98.0% (by HPLC, total nitrogen)

Supplier: Decon Laboratories Ltd
Warning

D5373-25G D5373-5G
TCD5373-25GEA 369 USD InStock
TCD5373-25G TCD5373-5G
1-Decyl-3-methylimidazoliumtetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
1-Decyl-3-methylimidazoliumtetrafluoroborate


Formula: C₁₄H₂₇BF₄N₂
Melting Pt: -4°C
MDL Number: MFCD09038853
CAS Number: 244193-56-4
Order Now
Specification Test Results
Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty