Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Formylhydrazine ≥97.0% (by HPLC)

Supplier: Decon Laboratories Ltd
Warning

F0667-100G F0667-25G
TCF0667-100GEA 168 USD InStock
TCF0667-100G TCF0667-25G
Formylhydrazine ≥97.0% (by HPLC)
Formylhydrazine


Formula: CH₄N₂O
Boiling Pt: 90°C
Storage Temperature: Refrigerator
MDL Number: MFCD00007589
CAS Number: 624-84-0
EINECS: 210-867-9
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 56.0 to 61.0 deg-C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty