Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(trifluoromethyl)phenol ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

B4492-1G B4492-5G
TCB4492-1GEA 77 USD InStock
TCB4492-1G TCB4492-5G
2-Bromo-4-(trifluoromethyl)phenol ≥98.0% (by GC, titration analysis)
2-Bromo-4-(trifluoromethyl)phenol


Formula: C₇H₄BrF₃O
MW: 241.01
MDL Number: MFCD06657954
CAS Number: 81107-97-3
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty