Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-fluorobenzonitrile ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd
Danger

C3103-1G C3103-5G
TCC3103-1GEA 41 USD InStock
TCC3103-1G TCC3103-5G
4-Chloro-3-fluorobenzonitrile ≥98.0% (by GC)
4-Chloro-3-fluorobenzonitrile


Formula: C₇H₃ClFN
MW: 155.56
MDL Number: MFCD00143291
CAS Number: 110888-15-8
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty