Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxybenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

H1510-1G H1510-5G
TCH1510-1GEA 41 USD InStock
TCH1510-1G TCH1510-5G
4-Hydroxybenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Hydroxybenzamidine hydrochloride


Formula: C₇H₉ClN₂O
MW: 172.61
MDL Number: MFCD00136405
CAS Number: 38148-63-9
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 221.0 to 225.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty