Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethyl-3-methylimidazolium diethylphosphate ≥96.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd
Danger

E1309-25G E1309-5G
TCE1309-25GEA 90 USD InStock
TCE1309-25G TCE1309-5G
1-Ethyl-3-methylimidazolium diethylphosphate ≥96.0% (by HPLC, titration analysis)
1-Ethyl-3-methylimidazolium diethylphosphate


Formula: C₁₀H₂₁N₂O₄P
MW: 264.26 g/mol
MDL Number: MFCD09953486
CAS Number: 848641-69-0
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Appearance Colorless to Yellow to Orange clear liquid
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty