Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Butyl-3-methylimidazolium methanesulfonate ≥98.0% (by HPLC, total nitrogen)

Supplier: Decon Laboratories Ltd
Danger

B3231-25G B3231-5G
TCB3231-25GEA 120 USD InStock
TCB3231-25G TCB3231-5G
1-Butyl-3-methylimidazolium methanesulfonate ≥98.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazolium methanesulphonate


Formula: C₉H₁₈N₂O₃S
MDL Number: MFCD06798173
CAS Number: 342789-81-5
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 74.0 to 78.0 deg-C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty