Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

F1094-1G F1094-5G
TCF1094-1GEA 85 USD InStock
TCF1094-1G TCF1094-5G
2-Fluoro-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Fluoro-5-pyridineboronic acid pinacol ester


Formula: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03412792
CAS Number: 444120-95-0
Order Now
Specification Test Results
Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 29.0 to 33.0 deg-C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty