Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahexylammonium hydrogen sulfate ≥98.0% (by total nitrogen and titration analysis)

Supplier: Decon Laboratories Ltd
Warning

T3495-25G T3495-5G
TCT3495-25GEA 292 USD InStock
TCT3495-25G TCT3495-5G
Tetrahexylammonium hydrogen sulfate ≥98.0% (by total nitrogen and titration analysis)
Tetrahexylammonium hydrogen sulfate


Formula: C₂₄H₅₃NO₄S
MW: 451.76
Melting Pt: 98°C
MDL Number: MFCD00037675
CAS Number: 32503-34-7
EINECS: 251-069-0
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty