Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecylphosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd
Danger

D4809-1G D4809-5G
TCD4809-1GEA 43 USD InStock
TCD4809-1G TCD4809-5G
Dodecylphosphonic acid ≥98.0% (by titrimetric analysis)
Dodecylphosphonic acid


Formula: C₁₂H₂₇O₃P
MW: 250.31 g/mol
MDL Number: MFCD00015832
CAS Number: 5137-70-2
EINECS: 225-897-8
UN: 3261
ADR: 8 III
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty