Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminofluoranthene ≥96.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

A2789-1G A2789-5G
TCA2789-1GEA 40 USD InStock
TCA2789-1G TCA2789-5G
3-Aminofluoranthene ≥96.0% (by GC, titration analysis)
3-Aminofluoranthene


Formula: C₁₆H₁₁N
MW: 217.2700 g/mol
MDL Number: MFCD00001185
CAS Number: 2693-46-1
EINECS: 220-263-7
UN: 3077
ADR: 9 III
Order Now
Specification Test Results
Appearance White to Amber powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 117.0 to 121.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty