Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Glutamic acid 5-ethyl ester ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

E1155-25G E1155-5G
TCE1155-25GEA 93 USD InStock
TCE1155-25G TCE1155-5G
L-Glutamic acid 5-ethyl ester ≥97.0% (by titrimetric analysis)
L-Glutamic acid 5-ethyl ester


Formula: C₇H₁₃NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00037217
CAS Number: 1119-33-1
EINECS: 214-274-6
Merck Index: 14,04470
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D +11.0 to +14.0 deg (C=1,H2O)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty