Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Acetamidoacetophenone ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd
Warning

A2821-25G A2821-5G
TCA2821-25GEA 252 USD InStock
TCA2821-25G TCA2821-5G
4'-Acetamidoacetophenone ≥98.0% (by GC)
4'-Acetamidoacetophenone


Formula: C₁₀H₁₁NO₂
MDL Number: MFCD00014965
CAS Number: 2719-21-3
EINECS: 220-320-6
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 169.0 to 173.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty