Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone ≥98.0% (by HPLC)

Supplier: Decon Laboratories Ltd

B5519-5G B5519-1G
76218-834EA 371 USD InStock
76218-834 76219-648
(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone ≥98.0% (by HPLC)
(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone


Formula: C₂₅H₂₄N₂O₂
MDL Number: MFCD00145260
CAS Number: 96293-17-3
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 98.0 to 102.0 deg-C
Specific rotation [a]20/D -136 to -142 deg (C=1,MeOH)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty