Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-hydroxybenzophenone ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd
Warning

C0186-25G C0186-5G
TCC0186-25GEA 144 USD InStock
TCC0186-25G TCC0186-5G
5-Chloro-2-hydroxybenzophenone ≥98.0% (by GC)
5-Chloro-2-hydroxybenzophenone


Formula: C₁₃H₉ClO₂
MDL Number: MFCD00020134
CAS Number: 85-19-8
EINECS: 201-592-5
Order Now
Specification Test Results
Purity (GC) min. 98.0 %
Melting point 95.0 to 99.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty