Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenylethylboronic acid (contains varying amounts of Anhydride)

Supplier: Decon Laboratories Ltd
Warning

P2476-1G P2476-5G
TCP2476-1GEA 29 USD InStock
TCP2476-1G TCP2476-5G
2-Phenylethylboronic acid (contains varying amounts of Anhydride)
2-Phenylethylboronic acid


Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
Melting Pt: 88°C
MDL Number: MFCD01631226
CAS Number: 34420-17-2
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 113.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty