Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-6-phenylpyridazine ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd
Warning

C3440-1G C3440-5G
TCC3440-1GEA 120 USD InStock
TCC3440-1G TCC3440-5G
3-Chloro-6-phenylpyridazine ≥98.0% (by GC)
3-Chloro-6-phenylpyridazine


Formula: C₁₀H₇ClN₂
MW: 190.6300 g/mol
MDL Number: MFCD00075253
CAS Number: 20375-65-9
EINECS: 243-772-6
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 157.0 to 161.0 deg-C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty