Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Imidazolecarboxylic acid ≥98.0% (by HPLC)

Supplier: Decon Laboratories Ltd
Warning

I1008-1G I1008-5G
TCI1008-1GEA 86 USD InStock
TCI1008-1G TCI1008-5G
2-Imidazolecarboxylic acid ≥98.0% (by HPLC)
2-Imidazolecarboxylic acid


Formula: C₄H₄N₂O₂
Melting Pt: 164°C
MDL Number: MFCD00270816
CAS Number: 16042-25-4
Order Now
Specification Test Results
Appearance White to Dark blue powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty