Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-amino-4-(trifluoromethyl)benzoate ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

M2905-1G M2905-5G
TCM2905-1GEA 66 USD InStock
TCM2905-1G TCM2905-5G
Methyl-2-amino-4-(trifluoromethyl)benzoate ≥98.0% (by GC, titration analysis)
Methyl-2-amino-4-(trifluoromethyl)benzoate


Formula: C₉H₈F₃NO₂
MDL Number: MFCD08543939
CAS Number: 61500-87-6
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 61.0 to 65.0 deg-C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty