Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-1,3,5-triazine ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

D4891-1G D4891-5G
TCD4891-1GEA 44 USD InStock
TCD4891-1G TCD4891-5G
2,4-Dichloro-1,3,5-triazine ≥98.0% (by GC, titration analysis)
2,4-Dichloro-1,3,5-triazine


Formula: C₃HCl₂N₃
MW: 149.9661 g/mol
Boiling Pt: 102°C
MDL Number: MFCD04115347
CAS Number: 2831-66-5
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 54.0 to 58.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty