Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

n-Pentylboronic acid (contains varying amounts of Anhydride)

Supplier: Decon Laboratories Ltd
Warning

P2267-25G P2267-5G
TCP2267-25GEA 298 USD InStock
TCP2267-25G TCP2267-5G
n-Pentylboronic acid (contains varying amounts of Anhydride)
n-Pentylboronic acid


Formula: C₅H₁₃BO₂
Melting Pt: 94°C
MDL Number: MFCD01074651
CAS Number: 4737-50-2
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 119.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty