Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Aminomethylquinoline ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

A2908-1G A2908-200MG
TCA2908-1GEA 260 USD InStock
TCA2908-1G TCA2908-200MG
6-Aminomethylquinoline ≥98.0% (by GC, titration analysis)
6-Aminomethylquinoline


Formula: C₁₀H₁₀N₂
Boiling Pt: 159°C
Density: 1.02
MDL Number: MFCD02853688
CAS Number: 99071-54-2
Order Now
Specification Test Results
Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty