Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzyloxy)acetaldehyde ≥95.0% (by GC) stabilised

Supplier: Decon Laboratories Ltd
Warning

Stabilization: Stabilised with catechol

B2481-1G B2481-5G
76219-354EA 59 USD InStock
76219-354 76219-356
2-(Benzyloxy)acetaldehyde ≥95.0% (by GC) stabilised
2-(Benzyloxy)acetaldehyde


Formula: C₉H₁₀O₂
MW: 150.177 g/mol
Boiling Pt: 120°C
Storage Temperature: Refrigerator
MDL Number: MFCD00191779
CAS Number: 60656-87-3
EINECS: 262-349-7
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty