Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mangostine ≥97.0% (by HPLC)

Supplier: Decon Laboratories Ltd
Warning

M2793-100MG M2793-10MG
TCM2793-100MGEA 511 USD InStock
TCM2793-100MG TCM2793-10MG
Mangostine ≥97.0% (by HPLC)
Mangostine


Formula: C₂₄H₂₆O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00135200
CAS Number: 6147-11-1
Order Now
Specification Test Results
Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 179.0 to 183.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty