Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridine-2-carboxamidoxime ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

P2590-25G P2590-5G
TCP2590-25GEA 159 USD InStock
TCP2590-25G TCP2590-5G
Pyridine-2-carboxamidoxime ≥98.0% (by HPLC, titration analysis)
Pyridine-2-carboxamidoxime


Formula: C₆H₇N₃O
MDL Number: MFCD00085159
CAS Number: 1772-01-6
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty