Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

M2717-1G M2717-200MG
TCM2717-1GEA 430 USD InStock
TCM2717-1G TCM2717-200MG
2-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole ≥98.0% (by GC, titration analysis)
2-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole


Formula: C₁₂H₁₄N₂
MDL Number: MFCD00460783
CAS Number: 5094-12-2
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 170.0 to 174.0 deg-C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty