Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(tert-Butyldimethylsilyl)thiazole ≥97.0% (by GC)

Supplier: Decon Laboratories Ltd
Warning

B4654-1G B4654-5G
TCB4654-1GEA 35.6 USD InStock
TCB4654-1G TCB4654-5G
2-(tert-Butyldimethylsilyl)thiazole ≥97.0% (by GC)
2-(tert-Butyldimethylsilyl)thiazole


Formula: C₉H₁₇NSSi
MDL Number: MFCD10699169
CAS Number: 137382-38-8
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty