Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoro-1H-indole-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

F0835-1G F0835-200MG
TCF0835-1GEA 427 USD InStock
TCF0835-1G TCF0835-200MG
6-Fluoro-1H-indole-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
6-Fluoro-1H-indole-2-carboxylic acid


Formula: C₉H₆FNO₂
MDL Number: MFCD01863162
CAS Number: 3093-97-8
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty