Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,3,3,3-Pentafluoropropylamine ≥97.0% (by GC)

Supplier: Decon Laboratories Ltd
Danger

P2281-1G P2281-5G
TCP2281-1GEA 72.2 USD InStock
TCP2281-1G TCP2281-5G
2,2,3,3,3-Pentafluoropropylamine ≥97.0% (by GC)
2,2,3,3,3-Pentafluoropropylamine


Formula: C₃H₄F₅N
MW: 149.06 g/mol
Boiling Pt: 50°C
Storage Temperature: Refrigerator
MDL Number: MFCD00042459
CAS Number: 422-03-7
EINECS: 207-010-6
UN: 2735
ADR: 8 III
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty