Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-n-Butyl-3-methylimidazolium hexafluoroantimonate ≥98.0% (by HPLC, total nitrogen)

Supplier: Decon Laboratories Ltd
Warning

B5724-25G B5724-5G
TCB5724-25GEA 87.98 USD InStock
TCB5724-25G TCB5724-5G
1-n-Butyl-3-methylimidazolium hexafluoroantimonate ≥98.0% (by HPLC, total nitrogen)
1-n-Butyl-3-methylimidazolium hexafluoroantimonate


Formula: C₈H₁₅F₆N₂Sb
MDL Number: MFCD07784453
CAS Number: 174645-81-9
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty