Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,8-Diiododibenzofuran ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

D4835-1G D4835-200MG
TCD4835-1GEA 299.2 USD InStock
TCD4835-1G TCD4835-200MG
2,8-Diiododibenzofuran ≥98.0% (by GC)
2,8-Diiododibenzofuran


Formula: C₁₂H₆I₂O
MDL Number: MFCD00094363
CAS Number: 5943-11-3
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 174.0 to 178.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty