Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-(3-Bromophenyl)-10-phenylanthracene ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

B4897-1G B4897-5G
TCB4897-1GEA 177 USD InStock
TCB4897-1G TCB4897-5G
9-(3-Bromophenyl)-10-phenylanthracene ≥98.0% (by GC)
9-(3-Bromophenyl)-10-phenylanthracene


Formula: C₂₆H₁₇Br
MDL Number: MFCD28100940
CAS Number: 1023674-80-7
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystaline
Purity (GC) min. 98.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty