Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tributylhexylphosphonium bromide ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd
Warning

T3840-25G T3840-5G
TCT3840-25GEA 333 USD InStock
TCT3840-25G TCT3840-5G
Tributylhexylphosphonium bromide ≥98.0% (by titrimetric analysis)
Tributylhexylphosphonium bromide


Formula: C₁₈H₄₀BrP
MDL Number: MFCD00134170
CAS Number: 105890-71-9
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty