Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,9-Diphenyl-9H-carbazole-3-boronic acid (contains varying amounts of Anhydride)

Supplier: Decon Laboratories Ltd
Warning

D4779-1G D4779-200MG
TCD4779-1GEA 240 USD InStock
TCD4779-1G TCD4779-200MG
6,9-Diphenyl-9H-carbazole-3-boronic acid (contains varying amounts of Anhydride)
6,9-Diphenyl-9H-carbazole-3-boronic acid


Formula: C₂₄H₁₈BNO₂
MDL Number: MFCD22123650
CAS Number: 1133058-06-6
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 105.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty